Anti-Itgb2 Rat Monoclonal Antibody [clone: C71/16] (PE (Phycoerythrin))
Catalog # PRSI98-655
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:integrin beta 2
- Antigen symbol:Itgb2
- Clonality:Monoclonal
- Clone:[C71/16]
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Form:Liquid
- Gene ID:16414
- Antigen synonyms:ITGB2|Integrin beta2
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃ and a stabilising agent
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,1 mg/ml
- Shipping temperature:20
- Immunogen:Cell membrane glycoproteins derived from mouse BW5147 T cell lymphoma cells
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Itgb2
Clonality: Monoclonal
Clone: [C71/16]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse