Anti-HSPA8 Rabbit Polyclonal Antibody
PRSI18-662
- Antibody type:Primary
- Antigen name:Heat shock 70kDa protein 8
- Antigen symbol:HSPA8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3312
- Antigen synonyms:HSP73|HSC70|constitutive heat shock protein 70|heat shock cognate protein|HSC71|heat shock 70 kDa protein 8|lipopolysaccharide-associated protein|heat shock 70kd protein 10|NIP71|HSPA10|N-myristoyltransferase inhibitor protein 71|LPS-associated protein 1|MGC131511|HSC54|MGC29929|heat shock 70kD protein 8|LAP|HSP71|HSPA8|71- kDa|heat shock cognate protein 54|uncharacterized bone marrow protein BM034
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Hsc70 (NP_006588.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: HSPA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat