Anti-ZNF268 Rabbit Polyclonal Antibody
PRSI14-693
- Antibody type:Primary
- Antigen name:zinc finger protein 268
- Antigen symbol:ZNF268
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10795
- Antigen synonyms:zinc finger protein 3|zinc finger protein 268|zinc finger protein HZF3|HZF3
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ZNF268 (NP_001159353.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ZNF268
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat