Anti-SEMG2 Rabbit Polyclonal Antibody
Vial
Catalog # PRSI14-648
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
Type: Primary
Antigen: SEMG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:semenogelin 2
- Cat. No.:PRSI14-648
- Reactivity:Human|Mouse
- Gene ID:6407
- Antigen synonyms:semenogelin-2|semenogelin II|SGII
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 383-582 of human SEMG2 (NP_002999.1)
- Conjugation:Unconjugated
- Antigen symbol:SEMG2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3



