Anti-ALKBH4 Rabbit Polyclonal Antibody
Vial
Catalog # PRSI14-530
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
Type: Primary
Antigen: ALKBH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:Alpha-ketoglutarate-dependent dioxygenase alkB homolog 4
- Cat. No.:PRSI14-530
- Gene ID:54784
- Antigen synonyms:Alkylated DNA repair protein alkB homolog 4|ALKBH4|11411-|Alpha-ketoglutarate-dependent dioxygenase alkB homolog 4|ABH4
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-302 of human ALKBH4 (NP_060091.1)
- Conjugation:Unconjugated
- Antigen symbol:ALKBH4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3



