Anti-USH1C Rabbit Polyclonal Antibody
Catalog # PRSI14-505
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Usher syndrome 1C (autosomal recessive, severe)
- Antigen symbol:USH1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10083
- Antigen synonyms:AIE-75|PDZ-73/NY-CO-38|deafness|HARMONIN|AIE75|Usher syndrome 1C (autosomal recessive|NY-CO-38|severe)|PDZ-73|DFNB18|PDZ-73 protein|USH1C|autosomal recessive 18|harmonin|NY-CO-37|ush1cpst|PDZ-45
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 264-533 of human USH1C (NP_001284693.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: USH1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat