Order Entry
Export
ContactUsLinkComponent
Anti-ATP6V1E2 Rabbit Polyclonal Antibody
Anti-ATP6V1E2 Rabbit Polyclonal Antibody
Catalog # PRSI14-313
Supplier:  ProSci Inc.
Anti-ATP6V1E2 Rabbit Polyclonal Antibody
Catalog # PRSI14-313
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATPase H+ transporting V1 subunit E2
  • Antigen symbol:
    ATP6V1E2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    90423
  • Antigen synonyms:
    ATP6EL2|VMA4|ATP6E1|lysosomal 31kDa|V-ATPase subunit E 2|ATP6V1EL2|vacuolar-type proton-translocating ATPase subunit E1|V1 subunit E2|V-type proton ATPase subunit E 2|vacuolar proton pump subunit E 2|testis secretory sperm-binding protein Li 235P|H+ transporting|ATPase
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at –20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 90-180 of human ATP6V1E2 (NP_542384.1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: ATP6V1E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat