Anti-TRMT61A Rabbit Polyclonal Antibody
Catalog # PRSI13-751
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:tRNA methyltransferase 61A
- Antigen symbol:TRMT61A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:115708
- Antigen synonyms:hTRM61|mRNA methyladenosine-N(1)-methyltransferase catalytic subunit TRMT61A|tRNA(m1A58)MTase subunit TRMT61A|tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61A|tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRM61|tRNA(m1A58)MTase subunit TRM61|GCD14|TRM61|tRNA(m1A58)-methyltransferase subunit TRMT61A|tRNA methyltransferase 61 homolog A|C14orf172|tRNA(m1A58)-methyltransferase subunit TRM61|Gcd14p|tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61A
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-289 of human TRMT61A (NP_689520.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TRMT61A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat