Anti-LGR5 Rabbit Polyclonal Antibody
PRSI13-711
- Antibody type:Primary
- Antigen name:Leucine-rich repeat-containing G-protein coupled receptor 5
- Antigen symbol:LGR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:8549
- Antigen synonyms:GPR67|GPR49|G-protein coupled receptor 49|LGR5|G-protein coupled receptor HG38|G-protein coupled receptor 67|Leucine-rich repeat-containing G-protein coupled receptor 5
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 824-907 of human LGR5 (NP_003658.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: LGR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat