Anti-MTAP Rabbit Polyclonal Antibody
PRSI13-684
- Antibody type:Primary
- Antigen name:methylthioadenosine phosphorylase
- Antigen symbol:MTAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4507
- Antigen synonyms:MSAP|MTA phosphorylase|epididymis secretory sperm binding protein|MTAPase|epididymis luminal protein 249|DMSMFH|BDMF|DMSFH|MeSAdo phosphorylase|5'-methylthioadenosine phosphorylase|HEL-249|S-methyl-5'-thioadenosine phosphorylase|LGMBF|c86fus
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human MTAP (NP_002442.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: MTAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat