Anti-SLC44A4 Rabbit Polyclonal Antibody
Catalog # PRSI13-647
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 44 member 4
- Antigen symbol:SLC44A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:80736
- Antigen synonyms:NG22|CTL4|testicular tissue protein Li 48|thiamine pyrophosphate transporter|C6orf29|DFNA72|choline transporter-like protein 4|hTPPT1|TPPT|thiamine pyrophosphate transporter 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 65-215 of human SLC44A4 (NP_079533.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC44A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse