Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE (Phycoerythrin)/Texas Red®)
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
: For research use only.
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- AATB100411S0
- AATB100411S1
- AATB100411S2
- Antigen symbol:
- CD4
- CD4
- CD4
- Antigen name:
- CD4 antigen
- CD4 antigen
- CD4 antigen
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Texas Red®
- PE (Phycoerythrin)/Texas Red®
- PE (Phycoerythrin)/Texas Red®
- Clone:
- RPA-T4
- RPA-T4
- RPA-T4
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
- 20