Order Entry
Export
ContactUsLinkComponent
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® A7)
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® A7)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (iFluor® A7)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • AATB100810S0
  • AATB100810S1
  • Antigen symbol:
  • CD8a
  • CD8a
  • Antigen name:
  • CD8a antigen
  • CD8a antigen
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® A7
  • iFluor® A7
  • Clone:
  • SK1
  • SK1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • 2...8 °C with minimised light exposure. Do not freeze.
  • 2...8 °C with minimised light exposure. Do not freeze.
  • Shipping temperature:
  • 20
  • 20

Specifications