Anti-LGR5 Rabbit Polyclonal Antibody
Catalog # PRSI23-394
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Leucine-rich repeat-containing G-protein coupled receptor 5
- Antigen symbol:LGR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8549
- Antigen synonyms:GPR67|GPR49|G-protein coupled receptor 49|LGR5|G-protein coupled receptor HG38|G-protein coupled receptor 67|Leucine-rich repeat-containing G-protein coupled receptor 5
- Storage buffer:PBS with 0,02% sodium azide, pH 7,3
- Storage temperature:Store at 4...730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human LGR5
- Purification:Affinity purification
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: LGR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat