Anti-NAA50 Rabbit Polyclonal Antibody
Catalog # PRSI22-907
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:N(alpha)-acetyltransferase 50, NatE catalytic subunit
- Antigen symbol:NAA50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:80218
- Antigen synonyms:NAT5|hNAT5|FLJ13194|hSAN|SAN|NAT13|MAK3|NAT5P|NAT13P
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-169 of human NAA50 (NP_079422.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NAA50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat