Order Entry
Export
ContactUsLinkComponent
Anti-UBASH3B Rabbit Polyclonal Antibody
Anti-UBASH3B Rabbit Polyclonal Antibody
  PRSI22-735
Anti-UBASH3B Rabbit Polyclonal Antibody
  PRSI22-735

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin associated and SH3 domain containing B
  • Antigen symbol:
    UBASH3B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    84959
  • Antigen synonyms:
    T-cell ubiquitin ligand 2|suppressor of T-cell receptor signaling 1|TULA-2|TULA2|cbl-interacting protein p7nm23-phosphorylated unknown substrate|SH3 domain-containing 70 kDa protein|STS1|Cbl-interacting protein Sts-1|tyrosine-protein phosphatase STS1/TULA2|STS-1|nm23-phosphorylated unknown substrate|p7ubiquitin-associated and SH3 domain-containing protein B
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human UBASH3B (NP_116262.2)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: UBASH3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat