Anti-WDR77 Rabbit Polyclonal Antibody
Catalog # PRSI22-729
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:WD repeat domain 77
- Antigen symbol:WDR77
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:79084
- Antigen synonyms:MEP50|WDR77|Methylosome protein 50|Androgen receptor cofactor p44|WD repeat domain 77|MEP-50|Nbla10071|HKMT1069|p44|RP11-552M11.3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human WDR77 (NP_077007.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: WDR77
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse