Anti-CSRNP1 Rabbit Polyclonal Antibody
Catalog # PRSI22-726
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Cysteine/serine-rich nuclear protein 1
- Antigen symbol:CSRNP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:64651
- Antigen synonyms:CSRNP1|TAIP3|CSRNP-1|TGF-beta-induced apoptosis protein 3|Cysteine/serine-rich nuclear protein 1|TAIP-3|AXUD1|Protein URAX1|Axin-1 up-regulated gene 1 protein
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CSRNP1 (NP_149016.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CSRNP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat