Anti-STAM2 Rabbit Polyclonal Antibody
Vial
PRSI22-666
Type: Primary
Antigen: STAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:Signal transducing adaptor molecule (SH3 domain and ITAM motif) 2
- Cat. No.:PRSI22-666
- Gene ID:10254
- Antigen synonyms:HGNC:11358|HBP|STAM2B|STAM-like protein containing SH3 and ITAM domains 2|signal transducing adaptor molecule (SH3 domain and ITAM motif) 2|DKFZp564C047|signal transducing adaptor molecule 2|STAM2A|Hbp|STAM2
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 376-525 of human STAM2 (NP_005834.4)
- Conjugation:Unconjugated
- Antigen symbol:STAM2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3



