Anti-GABBR2 Rabbit Polyclonal Antibody
Catalog # PRSI22-338
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid type B receptor subunit 2
- Antigen symbol:GABBR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:9568
- Antigen synonyms:GABA-B-R2|GABABR2|GABA-BR2|GABA-B receptor 2|GABBR2|Gamma-aminobutyric acid type B receptor subunit 2|Gb2|GPRC3B|G-protein coupled receptor 51|GPR51|HG20
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 60-310 of human GABBR2 (NP_005449.5)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GABBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat