Anti-MYOT Rabbit Polyclonal Antibody
Catalog # PRSI22-222
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Myotilin
- Antigen symbol:MYOT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:9499
- Antigen synonyms:myofibrillar titin-like Ig domains protein|myotilin|OTTHUMP00000223461|MYOT|LGMD1A|OTTHUMP00000223460|OTTHUMP00000159430|TTID|57 kDa cytoskeletal protein|titin immunoglobulin domain protein (myotilin)|LGMD1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 259-314 of human MYOT (NP_001129412.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MYOT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat