Anti-UBE3A Rabbit Polyclonal Antibody
Catalog # PRSI16-987
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin-protein ligase E3A
- Antigen symbol:UBE3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:7337
- Antigen synonyms:E6AP|Human papillomavirus E6-associated protein|UBE3A|Ubiquitin-protein ligase E3A|632-|EPVE6AP|Oncogenic protein-associated protein E6-AP|Renal carcinoma antigen NY-REN-54|E6AP ubiquitin-protein ligase|HPVE6A
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 300-590 of human UBE3A (NP_570853,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: UBE3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat