Anti-NECAB3 Rabbit Polyclonal Antibody
Catalog # PRSI16-078
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:N-terminal EF-hand calcium binding protein 3
- Antigen symbol:NECAB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:63941
- Antigen synonyms:Nek2-interacting protein 1|family A|SYTIP2|X11L-binding protein 51|dJ63M2.5|N-terminal EF-hand calcium-binding protein 3|EFCBP3|XB51|APBA2BP|member 2 binding protein|STIP3|neuronal calcium-binding protein NECAB3|synaptotagmin interacting protein STIP3|synaptotagmin interacting protein 2|amyloid beta A4 protein-binding family A member 2-binding protein|EF-hand calcium binding protein 3|amyloid beta (A4) precursor protein-binding|neuronal calcium-binding protein 3|NIP1|dJ63M2.4
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-362 of human NECAB3 (NP_112508,3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NECAB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat