Anti-GAP43 Rabbit Polyclonal Antibody
Catalog # PRSI22-172
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Growth associated protein 43
- Antigen symbol:GAP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2596
- Antigen synonyms:protein F1|axonal membrane protein GAP-43|B-5PP46|neuromodulin|nerve growth-related peptide GAP43|calmodulin-binding protein P-57|neural phosphoprotein B-5neuron growth-associated protein 43
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-238 of human GAP43 (NP_002036.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat