Anti-GC Rabbit Polyclonal Antibody
Catalog # PRSI19-807
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Group-specific component (vitamin D binding protein)
- Antigen symbol:GC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:2638
- Antigen synonyms:vitamin D-binding protein|VDBP|GRD3|group-specific component (vitamin D binding protein)|GC|vitamin D-binding protein/group specific component|DBP/GC|vitamin D-binding alpha-globulin|DBP|VDBG|HEL-S-51|vitamin D binding protein
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 324-493 of human GC (NP_001191236.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat