Anti-NPC2 Rabbit Polyclonal Antibody
Catalog # PRSI19-648
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Niemann-Pick disease, type C2
- Antigen symbol:NPC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10577
- Antigen synonyms:epididymal secretory protein|type C2|epididymal protein 1|EDDM1|HE1|Niemann-Pick disease|tissue-specific secretory protein|NPC2|epididymal secretory protein E1|MGC1333|NP-C2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 20-151 of human NPC2 (NP_006423.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse