Anti-REG1A Rabbit Polyclonal Antibody
Catalog # PRSI19-591
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Regenerating islet-derived 1 alpha
- Antigen symbol:REG1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5967
- Antigen synonyms:Pancreatic stone protein|PSP|REG1A|Lithostathine-1-alpha|PTP|ICRF|PSPS|Pancreatic thread protein|Protein-X|p19|Regenerating protein I alpha|Lithostathine 1 alpha|PSPS1|REG-1-alpha|REG
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 50-150 of human REG1A (NP_002900.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: REG1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat