Order Entry
Export
ContactUsLinkComponent
Anti-RACGAP1 Rabbit Polyclonal Antibody
Anti-RACGAP1 Rabbit Polyclonal Antibody
  PRSI19-569
Anti-RACGAP1 Rabbit Polyclonal Antibody
  PRSI19-569

 

  • Antibody type:
    Primary
  • Antigen name:
    Rac GTPase activating protein 1
  • Antigen symbol:
    RACGAP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    29127
  • Antigen synonyms:
    RACGAP1|MgcRacGAP|HsCYK-4|ID-GAP|CYK4
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 353-632 of human RACGAP1 (NP_001119576.1).
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: RACGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat