Anti-H3K14me1 (methylated) Rabbit Polyclonal Antibody
Catalog # PRSI19-558
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Histone cluster 3, H3
- Antigen symbol:H3K14me1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8290
- Antigen synonyms:H3.4|H3F3A|H3FT|H3t|H3/g
- Modification:Methylated
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human MonoMethyl-Histone H3-K14.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: H3K14me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat