Anti-NOP58 Rabbit Polyclonal Antibody
PRSI19-458
- Antibody type:Primary
- Antigen name:NOP58 ribonucleoprotein
- Antigen symbol:NOP58
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:51602
- Antigen synonyms:HSPC12NOP5|NOP58 ribonucleoprotein homolog|nucleolar protein 58|nucleolar protein 5|nucleolar protein NOP5/NOP58|NOP5/NOP58
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 360-529 of human NOP58 (NP_057018.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NOP58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human