Anti-UQCR10 Rabbit Polyclonal Antibody
Catalog # PRSI19-442
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ubiquinol-cytochrome c reductase, complex III subunit X
- Antigen symbol:UQCR10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:29796
- Antigen synonyms:UCRC|UCCR7.2|HSPC051|HSPC151|QCR9|HSPC119
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-63 of human UQCR10 (NP_037519.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: UQCR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat