Anti-AFF4 Rabbit Polyclonal Antibody
Catalog # PRSI19-432
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:AF4/FMR2 family member 4
- Antigen symbol:AFF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:27125
- Antigen synonyms:MCEF|AF4/FMR2 family member 4|AF5Q31|major CDK9 elongation factor-associated protein|ALL1-fused gene from chromosome 5q31 protein|CHOPS
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human AFF4 (NP_055238.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: AFF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse