Anti-FKBP5 Rabbit Polyclonal Antibody
Catalog # PRSI19-189
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:FKBP prolyl isomerase 5
- Antigen symbol:FKBP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2289
- Antigen synonyms:51 kDa FK506-binding protein|FK506 binding protein 5|peptidylprolyl cis-trans isomerase|54 kDa progesterone receptor-associated immunophilin|51 kDa FKBP|androgen-regulated protein 6|Ptg-1FKBP5|AIG61|peptidyl-prolyl cis-trans isomerase FKBP5|P54|HSP90-binding immunophilin|FKBP-51|FKBP54|FF1 antigen|T-cell FK506-binding protein|PPIase FKBP5|PPIase|rotamase
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 308-457 of human FKBP5 (NP_001139247.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: FKBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat