Anti-SFTPA1 Rabbit Polyclonal Antibody
Catalog # PRSI18-957
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Surfactant protein A1
- Antigen symbol:SFTPA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:653509
- Antigen synonyms:pulmonary-associated protein A1B|OTTHUMP00000019930|surfactant protein A1 variant AD' 6A|SFTPA1B|surfactant protein A1 variant ACD' 6A3|surfactant protein A1 variant AD' 6A3|FLJ51913|surfactant|OTTHUMP00000019928|pulmonary surfactant-associated protein A1|pulmonary-associated protein A1A|surfactant protein A1 variant AB'D' 6A2|collectin-4|SFTPA1|FLJ61144|surfactant protein A1 variant AD' 6A2|FLJ99559|surfactant protein A1|MGC198590|SPA1|SP-A|FLJ77898|SFTP1|PSAP|PSP-A|surfactant protein A1 variant AD' 6A4|SFTPA|FLJ50593|surfactant protein A1 variant AB'D' 6A3|COLEC4|SPA|SP-A1|FLJ79095|surfactant protein A1 variant ACD' 6A4|OTTHUMP00000019931|alveolar proteinosis protein|surfactant protein A1 variant ACD' 6A2|PSPA|surfactant protein A1 variant AB'D' 6A4|35 kDa pulmonary surfactant-associated protein|OTTHUMP00000019929|MGC133365|surfactant protein A1B
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human SFTPA1.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SFTPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse