Anti-GPRC6A Rabbit Polyclonal Antibody
PRSI60-142
New Product
- Antibody type:Primary
- Antigen name:G-protein coupled receptor family C group 6 member A
- Antigen symbol:GPRC6A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:489 - 517 aa
- Form:Liquid
- Gene ID:222545
- Modification:None
- Molecular weight:105 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This GPRC6A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 489 - 517 amino acids from the Central region of human GPRC6A.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: GPRC6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 489-517
Host: Rabbit
Isotype: Ig
Reactivity: Human