Anti-VPS54 Rabbit Polyclonal Antibody
Catalog # PRSI56-744
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Vacuolar protein sorting-associated protein 54
- Antigen symbol:VPS54
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Epitope:Amino acid range = 465-491
- Form:Liquid
- Gene ID:Q9P1Q0
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:111 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,35 mg/ml
- Shipping temperature:4 °C
- Immunogen:This VPS54 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 465-491 amino acids from the Central region of human VPS54
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: VPS54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Mouse