Anti-GPR34 Rabbit Polyclonal Antibody
PRSI56-686
- Antibody type:Primary
- Antigen name:Probable G-protein coupled receptor 34
- Antigen symbol:GPR34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acid range = 232-261
- Form:Liquid
- Gene ID:Q9UPC5
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:44 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This GPR34 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 232-261 amino acids from the Central region of human GPR34
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
G protein-coupled receptors (GPCRs), such as GPR34, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
Type: Primary
Antigen: GPR34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human