Anti-EIF2B1 Rabbit Polyclonal Antibody
Catalog # PRSI56-411
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B (EIF2B), a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
- For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: EIF2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- Modification:None
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Antibody type:Primary
- Epitope:Amino acid range = 136-163
- Form:Liquid
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Host:Rabbit
- Concentration:0,5 mg/ml
- Molecular weight:34 kDa
- Shipping temperature:4 °C
- Antigen name:Translation initiation factor eIF-2B subunit alpha
- Cat. No.:PRSI56-411
- Gene ID:Q14232
- Immunogen:This EIF2B1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 136-163 amino acids from the Central region of human EIF2B1
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:EIF2B1
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide



