Anti-FCER1A Mouse Monoclonal Antibody [clone: AER-37 (CRA1)] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-171
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
- Antigen symbol:FCER1A
- Clonality:Monoclonal
- Clone:AER-37 (CRA1)
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P12319
- Antigen synonyms:FCER1A
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The AER-37 (CRA1) antibody binds to the FC epsilon receptor I alpha subunit (FceR1a), a 50 kDa transmembrane protein. It is part of a tetrameric receptor complex composed from one alpha subunit, one beta subunit, and two gamma subunits. The FceR1a binds and is upregulated by the presence of IgE. It plays a role in prompting the allergic reactions mediated by IgE.
Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: AER-37 (CRA1)
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human