Order Entry
Export
ContactUsLinkComponent
Anti-Jag1 Hamster Monoclonal Antibody [clone: HMJ1-29] (PE (Phycoerythrin))
Anti-Jag1 Hamster Monoclonal Antibody [clone: HMJ1-29] (PE (Phycoerythrin))
Catalog # PRSI76-994
Supplier:  ProSci Inc.
Anti-Jag1 Hamster Monoclonal Antibody [clone: HMJ1-29] (PE (Phycoerythrin))
Catalog # PRSI76-994
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    jagged 1
  • Antigen symbol:
    Jag1
  • Clonality:
    Monoclonal
  • Clone:
    HMJ1-29
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Hamster
  • Isotype:
    IgG
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    Q9QXX0
  • Antigen synonyms:
    Jag1
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,1 mg

Specifications

About this item

The HMJ1-29 monoclonal antibody reacts with mouse and rat CD339 (Jagged 1), a type 1 transmembrane Notch receptor ligand. It is expressed by macrophages and certain stromal, epithelial, dendritic, and thymic lymphoid cells. It has been reported to be play a role in Treg induction, Th2 polarization, neurogenesis, and cardiovascular development.

Type: Primary
Antigen: Jag1
Clonality: Monoclonal
Clone: HMJ1-29
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse