Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9] (PE (Phycoerythrin))
PRSI76-146
- Antibody type:Primary
- Antigen name:integrin, alpha X (complement component 3 receptor 4 subunit)
- Antigen symbol:ITGAX
- Clonality:Monoclonal
- Clone:3.9
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P20702
- Antigen synonyms:ITGAX
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
The 3.9 monoclonal antibody specifically binds to the human adhesion glycoprotein CD11c, a 150 kDa integrin alpha chain also known as integrin alpha X. It is expressed on macrophages, granulocytes, monocytes, dendritic cells, natural killer cells, and subsets of B and T lymphocytes. The CD11c/CD18 complex associates with the iC3b, fibrinogen and ICAM-1 and has an important function in leukocyte adhesion.
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 3.9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human