Anti-SELL Mouse Monoclonal Antibody [clone: DREG-56]
Catalog # PRSI76-211
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:selectin L
- Antigen symbol:SELL
- Clonality:Monoclonal
- Clone:DREG-56
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Cross adsorption:No
- Format:purified antibody
- Form:liquid
- Gene ID:P14151
- Antigen synonyms:SELL
- Storage buffer:Phosphate-buffered aqueous solution, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Tested applications:Functional
- Purification:The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0,01 EU/µg of the protein.
- Pk:0,05 mg
Specifications
About this item
The DREG-56 monoclonal antibody specifically reacts with human CD62L, also known as L-selectin and LECAM-1. It mediates the migration of lymphocytes to the site of inflammation and their return to the peripheral lymphoid tissues and to the HEV (high endothelial venules). CD62L is expressed on monocytes, NK cells, neutrophils, granulocytes, and subsets of T and B cells. It is reported that the DREG-56 antibody inhibits the binding of lymphocytes to HEV tissue in frozen peripheral tissue.
Type: Primary
Antigen: SELL
Clonality: Monoclonal
Clone: DREG-56
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human