Order Entry
Export
ContactUsLinkComponent
Anti-RNA-directed RNA polymerase Rabbit Polyclonal Antibody
Anti-RNA-directed RNA polymerase Rabbit Polyclonal Antibody
Catalog # PRSI11-035
Supplier:  ProSci Inc.
Anti-RNA-directed RNA polymerase Rabbit Polyclonal Antibody
Catalog # PRSI11-035
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Form:
    Liquid
  • Antigen synonyms:
    M|Membrane protein|Matrix glycoprotein|E1 glycoprotein
  • Modification:
    None
  • Storage buffer:
    PBS, pH 7,4, containing 0,05% proclin300, 50% glycerol
  • Molecular weight:
    25 kDa
  • Storage temperature:
    Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2…8 °C for 1 - 2 weeks. Store at –20 °C for 12 months. Store at –80 °C for long term storage.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    MADSNGTITVEELKKLLEQWNLVIGFLFLTWICLLQFAYANRNRFLYIIKLIFLWLLWPVTLACFVLAAVYRINWITGGIAIAMACLVGLMWLSYFIASFRLFARTRSMWSFNPETNILLNVPLHGTILTRPLLESELVIGAVILRGHLRIAGHHLGRCDIKDLPKEITVATSRTLSYYKLGASQRVAGDSGFAAYSRYRIGNYKLNTDHSSSSDNIALLVQ
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus