Anti-NSD1 Rabbit Polyclonal Antibody
Catalog # PRSI24-011
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:nuclear receptor binding SET domain protein 1
- Antigen symbol:NSD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:64324
- Antigen synonyms:STO|ARA267|androgen receptor-associated protein of 267 kDa|H3-K36-HMTase|nuclear receptor SET domain-containing protein 1|androgen receptor-associated coregulator 267|histone-lysine N-methyltransferase|lysine N-methyltransferase 3B|H3 lysine-36 and H4 lysine-20 specific|androgen receptor coactivator 267 kDa protein|SOTOS1|SOTOS|KMT3B|NR-binding SET domain-containing protein|truncated nuclear receptor binding SET domain protein 1|nuclear receptor-binding SET domain-containing protein 1|H4-K20-HMTase
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 60-260 of human NSD1 (NP_071900.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NSD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse