Anti-SMU1 Rabbit Polyclonal Antibody
Catalog # PRSI23-794
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:SMU1 DNA replication regulator and spliceosomal factor
- Antigen symbol:SMU1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:55234
- Antigen synonyms:fSAP57|smu-1 suppressor of mec-8 and unc-52 protein homolog|SMU-1|functional spliceosome-associated protein 57|DNA replication regulator and spliceosomal factor|smu-1 suppressor of mec-8 and unc-52 homolog|brain-enriched WD-repeat protein|WD40 repeat-containing protein SMU1|BWD
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human SMU1 (NP_060695.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SMU1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat