Anti-DDT Rabbit Polyclonal Antibody
Vial
Catalog # PRSI23-731
Supplier: ProSci Inc.
Jump to:
Product Details & Documents
Type: Primary
Antigen: DDT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Pack type:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:None
- Purification:Affinity purification.
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Concentration:Batch dependent
- Shipping temperature:20
- Antigen name:D-dopachrome tautomerase
- Cat. No.:PRSI23-731
- Reactivity:Human|Rat|Mouse
- Gene ID:1652
- Antigen synonyms:D-dopachrome decarboxylase|MIF2|D-DT|MIF-2|DDCT|phenylpyruvate tautomerase II
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human DDT (NP_001346.1)
- Conjugation:Unconjugated
- Antigen symbol:DDT
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3



