Anti-CRNN Rabbit Polyclonal Antibody
Catalog # PRSI23-719
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:cornulin
- Antigen symbol:CRNN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:49860
- Antigen synonyms:cornulin|C1orf1DRC1|SEP53|53 kDa putative calcium-binding protein|PDRC1|squamous epithelial heat shock protein 53|58 kDa heat shock protein|53 kDa squamous epithelial-induced stress protein|tumor-related protein
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 396-495 of human CRNN (NP_057274.1)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CRNN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse