Order Entry
Export
ContactUsLinkComponent
Anti-SMEK1 Rabbit Polyclonal Antibody
Anti-SMEK1 Rabbit Polyclonal Antibody
Catalog # PRSI23-626
Supplier:  ProSci Inc.
Anti-SMEK1 Rabbit Polyclonal Antibody
Catalog # PRSI23-626
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SMEK homolog 1, suppressor of mek1 (Dictyostelium)
  • Antigen symbol:
    SMEK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    55671
  • Antigen synonyms:
    SMEK homolog 1|smk-1|FLFL1|PPP4R3A|MSTP033|KIAA2010|suppressor of mek1 (Dictyostelium)|PP4R3A|smk1|suppressor of mek1
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 395-594 of human SMEK1 (NP_001271210.1)
  • Purification:
    Affinity purification.
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: SMEK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat