Anti-Vsir Recombinant Antibody [Clone: MH5A]
PRSI24-582
- Antibody type:Primary
- Antigen name:V-set immunoregulatory receptor
- Antigen symbol:Vsir
- Clonality:Recombinant
- Clone:MH5A
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG lambda
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:74048
- Antigen synonyms:B7-H5|C10orf54|GI24|PP2135|DD1alpha|SISP1|chromosome 10 open reading frame 54|V-set immunoregulatory receptor|VISR|B7H5|VISTA
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:The antibody was generated by immunising Armenian hamsters with mPD-1HIg
- Tested applications:Blocking, therapeutics
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: V-set immunoregulatory receptor
Clonality: Recombinant
Clone: MH5A
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: Mouse