Anti-SELL Recombinant Antibody [Clone: DREG-56]
Catalog # PRSI24-440
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:selectin L
- Antigen symbol:SELL
- Clonality:Recombinant
- Clone:DREG-56
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Chimpanzee,Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:6402
- Antigen synonyms:LAM-1|LECAM1|Lymph node homing receptor|Lymphocyte surface MEL-14 antigen|MEL-14|Leukocyte adhesion molecule 1|Ly-22|LECAM-1|CD62 antigen-like family member L|Lymphocyte antigen 22|L-selectin|Leukocyte-endothelial cell adhesion molecule 1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was produced by immunizing mice with the concentrated supernatant generated after centrifugation of human peripheral blood leukocytes
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: selectin L
Clonality: Recombinant
Clone: DREG-56
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human;Chimpanzee;Bovine